Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxyethyl)trimethylammonium dimethyl phosphate, 95%, Thermo Scientific™

Catalog No. aah2760906
Click to view available options
Quantity:
5g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 118978-98-6
Molecular Formula C7H20NO5P
Linear Formula (HOCH2CH2)N(CH3)3·(CH3O)2PO2
Quantity 5g
MDL Number MFCD09953490
Synonym choline dimethyl phosphate, 2-hydroxyethyltrimethyl-ammonium dimethylphosphate, 2-hydroxyethyl trimethylammonium dimethyl phosphate, 2-hydroxyethyl trimethylammonium dimethylphosphate
InChI Key MCZQEJXSCZNROZ-UHFFFAOYSA-M
SMILES COP([O-])(=O)OC.C[N+](C)(C)CCO
IUPAC Name dimethyl phosphate;2-hydroxyethyl(trimethyl)azanium
Molecular Weight (g/mol) 229.21
PubChem CID 87076875
Formula Weight 229.21
Percent Purity 95%
Chemical Name or Material (2-Hydroxyethyl)trimethylammonium dimethyl phosphate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.