Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxyflavanone 98.0+%, TCI America™
SDP

Catalog No. h10241g
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 17348-76-4
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.26
MDL Number MFCD00017702
InChI Key KZKWCKFDCPVDFJ-UHFFFAOYNA-N
PubChem CID 176925
IUPAC Name 2-(2-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
SMILES OC1=CC=CC=C1C1CC(=O)C2=CC=CC=C2O1

Specifications

Melting Point 166°C
Quantity 1 g
Formula Weight 240.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxyflavanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.