Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 2-hydroxyisobutyryl-HIST1H2AG (Lys5) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121365 Supplier Invitrogen™ Supplier No. PA5121365
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody tested in Indirect ELISA.

Histones are nuclear proteins that play an important role in maintaining the structure of the chromosome in eukaryotes. The nucleosome is made up of around 146bp of DNA, which enfolds around a histone octomer unit comprising of four major histones, H2A, H2B, H3 and H4. An important role of H2A is to ensure proper assembly or functioning of the centrometre. They also play an important role in gene regulation. The protein can be modified by acetylation and phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 2-hydroxyisobutyryl-HIST1H2AG (Lys5)
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene H2AC11
Gene Accession No. P0C0S8
Gene Alias H2A histone family, member P; H2A.1; H2A.1b; H2A/d; H2A/p; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; H2AFC; H2AFD; H2AFI; H2AFN; H2AFP; H2AG; H2ak; HIST1H2AG; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; histone 1, H2ag; histone cluster 1, H2ag; histone H2A type 1; histone H2A/e; histone H2A/p; Histone H2A/ptl; pH2A/f
Gene Symbols H2AC11, HIST1H2AG
Host Species Rabbit
Immunogen Synthesized peptide derived from Human Histone H2A type 1 protein (2-14aa).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8329, 8969
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.