Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxypropiophenone 97.0+%, TCI America™
SDP

Catalog No. H0298500G
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 610-99-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002220
InChI Key KDUWXMIHHIVXER-UHFFFAOYSA-N
Synonym 2'-hydroxypropiophenone, 1-2-hydroxyphenyl propan-1-one, 2-propionylphenol, 1-propanone, 1-2-hydroxyphenyl, o-hydroxypropiophenone, o-propiophenol, propiophenone, 2'-hydroxy, ortho-hydroxypropiophenone, 1-propanone,1-2-hydroxyphenyl, 2-hydroxypropiophenone
PubChem CID 69133
IUPAC Name 1-(2-hydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 150°C
Quantity 500 g
Formula Weight 150.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Hydroxypropiophenone
EINECSNumber (3)-2945
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.