Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxypyridine-N-oxide, 97%

Catalog No. ac243790250
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 13161-30-3 | C5H5NO2 | 111.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13161-30-3
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
InChI Key SNUSZUYTMHKCPM-UHFFFAOYSA-N
Synonym 2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
PubChem CID 69975
IUPAC Name 1-hydroxypyridin-2-one
SMILES C1=CC(=O)N(C=C1)O

Specifications

Melting Point 147.0°C to 152.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 2-Hydroxypyridine-N-oxide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 236-100-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.