Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyquinoline, 99%, Thermo Scientific™

Catalog No. AAB2306406
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-31-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00006743
InChI Key LISFMEBWQUVKPJ-UHFFFAOYSA-N
Synonym 2-hydroxyquinoline, 2-quinolinol, quinolin-2-ol, carbostyril, 2 1h-quinolinone, 2-quinolone, quinolin-2 1h-one, quinolinol, quinolinone, alpha-quinolone
PubChem CID 6038
ChEBI CHEBI:18289
IUPAC Name 1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)N2

Specifications

Melting Point 196°C to 199°C
Quantity 5 g
Beilstein 2855
Merck Index 14,1825
Formula Weight 145.16
Percent Purity 99%
Chemical Name or Material 2-Hydroxyquinoline
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-420-6
RTECSNumber FG7175000
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.