Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxysaclofen, Thermo Scientific Chemicals

Catalog No. AAJ63588MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63588MF 50 mg
AAJ63588MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63588MF Supplier Thermo Scientific Chemicals Supplier No. J63588MF

Selective antagonist at GABA-B receptors

Selective antagonist at GABA-B receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117354-64-0
Molecular Formula C9H12ClNO4S
Molecular Weight (g/mol) 265.708
MDL Number MFCD00069282
InChI Key WBSMZVIMANOCNX-UHFFFAOYSA-N
Synonym 2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid
PubChem CID 1564
IUPAC Name 3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid
SMILES C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl

Specifications

Melting Point 268°C to 271°C
Color White
Quantity 50 mg
Physical Form Solid
UN Number UN1759
Solubility Information Soluble in water at 1mg/ml; Also soluble in DMSO,0;1M HCl and methanol
Formula Weight 265.71
Chemical Name or Material 2-Hydroxysaclofen
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
RTECSNumber DA0359200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.