Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Imidazolidinone hemihydrate, 98+%

Catalog No. p-7023778
Click to view available options
Quantity:
250 g
1000 g

CAS: 121325-67-5 | C6H14N4O3 | 190.20 g/mol

2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121325-67-5
Molecular Formula C6H14N4O3
Molecular Weight (g/mol) 190.20
MDL Number MFCD02662355
InChI Key KPRJGGOOWATRNT-UHFFFAOYSA-N
Synonym imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrate
PubChem CID 2723651
IUPAC Name imidazolidin-2-one;hydrate
SMILES O.O=C1NCCN1.O=C1NCCN1

Specifications

Melting Point 55°C
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 95.10 (86.09 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 2-Imidazolidinone hemihydrate
Hazard Category H319-H361f
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.