Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodo-1,1,1-trifluoroethane, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
353-83-3
C2H2F3I
209.94
MFCD00001082
RKOUFQLNMRAACI-UHFFFAOYSA-N
2-iodo-1,1,1-trifluoroethane,2,2,2-trifluoroethyl iodide,trifluoroethyl iodide,ethane, 1,1,1-trifluoro-2-iodo,1-iodo-2,2,2-trifluoroethane,2,2,2-trifluoroiodoethane,cf3ch2i,unii-ca0j38we9z,1,1,1-trifluoroiodoethane,1,1,1-trifluoro-2-iodo-ethane
67709
1,1,1-trifluoro-2-iodoethane
C(C(F)(F)F)I
353-83-3
C2H2F3I
209.94
MFCD00001082
RKOUFQLNMRAACI-UHFFFAOYSA-N
2-iodo-1,1,1-trifluoroethane,2,2,2-trifluoroethyl iodide,trifluoroethyl iodide,ethane, 1,1,1-trifluoro-2-iodo,1-iodo-2,2,2-trifluoroethane,2,2,2-trifluoroiodoethane,cf3ch2i,unii-ca0j38we9z,1,1,1-trifluoroiodoethane,1,1,1-trifluoro-2-iodo-ethane
67709
1,1,1-trifluoro-2-iodoethane
C(C(F)(F)F)I
Specific Gravity 2.13
Density 2.1300g/mL
Refractive Index 1.4000 to 1.4020
Packaging Glass bottle
Boiling Point 53.0°C to 55.0°C
Quantity 10mL
Infrared Spectrum Authentic
Color Colorless to Pink
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula ICH2CF3
Solubility Information Solubility in water: insoluble
Formula Weight 209.94
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Iodo-1, 1, 1-trifluoroethane, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-541-0

TSCA : TSCA