Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodo-4-nitrotoluene, 97%

Catalog No. aab2149106
Click to view available options
Quantity:
5 g
25 g

CAS: 7745-92-8 | C7H6INO2 | 263.03 g/mol

2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7745-92-8
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD00024329
InChI Key BUQSRXQJUZTIEW-UHFFFAOYSA-N
Synonym 2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821
PubChem CID 82188
IUPAC Name 2-iodo-1-methyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1I)[N+]([O-])=O

Specifications

Quantity 5 g
Sensitivity Light sensitive
Solubility Information Soluble in toluene.
Formula Weight 263.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-4-nitrotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-808-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.