Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodo-9,9-dimethylfluorene 98.0+%, TCI America™
SDP

Catalog No. I087525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I087525G 25 g
I08755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I087525G Supplier TCI America Supplier No. I087525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 144981-85-1
Molecular Formula C15H13I
Molecular Weight (g/mol) 320.17
MDL Number MFCD12024272
InChI Key DVLSJPCXPNKPRJ-UHFFFAOYSA-N
PubChem CID 11023630
IUPAC Name 2-iodo-9,9-dimethyl-9H-fluorene
SMILES CC1(C)C2=CC=CC=C2C2=C1C=C(I)C=C2

Specifications

Melting Point 66°C
Color Yellow
Quantity 25 g
Formula Weight 320.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-9,9-dimethylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.