Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodoaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
615-43-0
C6H6IN
219.02
MFCD00007680
UBPDKIDWEADHPP-UHFFFAOYSA-N
o-iodoaniline,benzenamine, 2-iodo,2-iodo aniline,2-iodobenzenamine,o-aminoiodobenzene,aniline, o-iodo,aniline, 2-iodo,benzenamine, iodo,2-iodo-phenylamine,2-iodophenylamine
11995
2-iodoaniline
C1=CC=C(C(=C1)N)I
615-43-0
C6H6IN
219.02
MFCD00007680
UBPDKIDWEADHPP-UHFFFAOYSA-N
o-iodoaniline,benzenamine, 2-iodo,2-iodo aniline,2-iodobenzenamine,o-aminoiodobenzene,aniline, o-iodo,aniline, 2-iodo,benzenamine, iodo,2-iodo-phenylamine,2-iodophenylamine
11995
2-iodoaniline
C1=CC=C(C(=C1)N)I
Flash Point >110°C
Quantity 25g
Color Brown to Yellow
Packaging Glass bottle
Melting Point 53°C to 58°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 12, 669
Solubility Information Solubility in water: slightly soluble
Formula Weight 219.02
Physical Form Crystalline Needles
Percent Purity ≥97.5%
Chemical Name or Material 2-Iodoaniline
Show More Show Less

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Harmful in contact with skin. May cause respiratory irritation. Harmful if inhaled.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning