Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzaldehyde, 98%

Catalog No. p-2690069
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC368170010

Supplier: Thermo Scientific Chemicals 368170010

Only null left
Add to Cart

CAS: 26260-02-6 | C7H5IO | 232.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26260-02-6
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039570
InChI Key WWKKTHALZAYYAI-UHFFFAOYSA-N
Synonym benzaldehyde, 2-iodo, o-iodobenzaldehyde, iodobenzaldehyde, benzaldehyde,iodo, 2-iodobenzenealdehyde, pubchem3914, acmc-209upi, 2-iodobenzaldehyde, ksc203m8l
PubChem CID 643439
IUPAC Name 2-iodobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34.0°C to 38.0°C
Color White to Beige
Boiling Point 129.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4CHO
Quantity 1 g
Formula Weight 232.02
Percent Purity 98%
Physical Form Low Melting Crystalline Mass or Powder
Chemical Name or Material 2-Iodobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.