Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodohippuric Acid 99.0+%, TCI America™
SDP

Catalog No. I044625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I044625G 25 g
I04465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I044625G Supplier TCI America Supplier No. I044625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 147-58-0
Molecular Formula C9H8INO3
Molecular Weight (g/mol) 305.071
MDL Number MFCD00002694
InChI Key CORFWQGVBFFZHF-UHFFFAOYSA-N
Synonym N-(2-Iodobenzoyl)glycine
PubChem CID 8614
IUPAC Name 2-[(2-iodobenzoyl)amino]acetic acid
SMILES C1=CC=C(C(=C1)C(=O)NCC(=O)O)I

Specifications

Melting Point 173°C
Color White
Quantity 25 g
Formula Weight 305.07
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodohippuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.