Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodothiophene, 98+%

Catalog No. ac187880100
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle

CAS: 3437-95-4 | C4H3IS | 210.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3437-95-4
Molecular Formula C4H3IS
Molecular Weight (g/mol) 210.03
MDL Number MFCD00005424
InChI Key ROIMNSWDOJCBFR-UHFFFAOYSA-N
Synonym thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene
PubChem CID 18921
IUPAC Name 2-iodothiophene
SMILES C1=CSC(=C1)I

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -40.0°C
Color Brown-Red to Green-Yellow
Density 1.9000g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.6510 to 1.6530
Quantity 10 mL
Beilstein 17, 34
Specific Gravity 1.9
Formula Weight 210.03
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Iodothiophene, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 222-342-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.