Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodothiophene, 98%, stab. with copper

Catalog No. p-7020816
Click to view available options
Quantity:
25 g
100 g

CAS: 3437-95-4 | C4H3IS | 210.032 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3437-95-4
Molecular Formula C4H3IS
Molecular Weight (g/mol) 210.032
MDL Number MFCD00005424
InChI Key ROIMNSWDOJCBFR-UHFFFAOYSA-N
Synonym thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene
PubChem CID 18921
IUPAC Name 2-iodothiophene
SMILES C1=CSC(=C1)I

Specifications

Melting Point -40°C
Density 2.064
Boiling Point 182°C
Flash Point 71°C (159°F)
Refractive Index 1.652
Quantity 25 g
Beilstein 104666
Sensitivity Light sensitive
Formula Weight 210.04
Percent Purity 98%
Chemical Name or Material 2-Iodothiophene, stabilized with copper
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H317-H318-H335
Hazard Statement GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
EINECSNumber 222-342-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.