Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodotoluene 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
615-37-2
C7H7I
218.04
MFCD00001042
RINOYHWVBUKAQE-UHFFFAOYSA-N
2-iodotoluene,o-iodotoluene,o-tolyl iodide,benzene, 1-iodo-2-methyl,o-methyliodobenzene,toluene, o-iodo,iodotoluene,ortho-iodotoluene,2-methyliodobenzene,unii-8ok4h85t07
5128
CHEBI:39813
1-iodo-2-methylbenzene
CC1=CC=CC=C1I
615-37-2
C7H7I
218.04
MFCD00001042
RINOYHWVBUKAQE-UHFFFAOYSA-N
2-iodotoluene,o-iodotoluene,o-tolyl iodide,benzene, 1-iodo-2-methyl,o-methyliodobenzene,toluene, o-iodo,iodotoluene,ortho-iodotoluene,2-methyliodobenzene,unii-8ok4h85t07
5128
CHEBI:39813
1-iodo-2-methylbenzene
CC1=CC=CC=C1I
Boiling Point 211°C
Flash Point 90°C
Specific Gravity 1.71
Infrared Spectrum Authentic
Density 1.7100g/mL
Refractive Index 1.6080 to 1.61
Color Orange to Yellow
Quantity 25mL
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4I
Beilstein 05, 310
Formula Weight 218.04
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Iodotoluene, 98%
Show More Show Less