Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodotriphenylene 98.0+%, TCI America™
SDP

Catalog No. I09781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09781G 1 g
I0978200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09781G Supplier TCI America Supplier No. I09781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1228778-59-3
Molecular Formula C18H11I
Molecular Weight (g/mol) 354.19
InChI Key ZMZNYRZSJDYTIO-UHFFFAOYSA-N
PubChem CID 88955426
IUPAC Name 2-iodotriphenylene
SMILES C1=CC=C2C(=C1)C3=C(C=C(C=C3)I)C4=CC=CC=C24

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 1 g
Formula Weight 354.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodotriphenylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.