Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isobutoxynaphthalene 98.0+%, TCI America™
SDP

Catalog No. N043325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N043325G 25 g
N0433500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N043325G Supplier TCI America Supplier No. N043325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2173-57-1
Molecular Formula C14H16O
Molecular Weight (g/mol) 200.28
MDL Number MFCD00046467
InChI Key XOHIHZHSDMWWMS-UHFFFAOYSA-N
Synonym Isobutyl 2-Naphthyl Ether
PubChem CID 16582
IUPAC Name 2-(2-methylpropoxy)naphthalene
SMILES CC(C)COC1=CC=C2C=CC=CC2=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 200.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Isobutoxynaphthalene
EINECSNumber (4)-0362
RTECSNumber KO1255000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.