Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers) 95.0+%, TCI America™
SDP

Catalog No. i073725g
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 65894-83-9
Molecular Formula C9H17NS
Molecular Weight (g/mol) 171.30
MDL Number MFCD01680822
InChI Key FDOISHJOXPONIV-UHFFFAOYNA-N
Synonym 4,5-dimethyl-2-isobutyl-3-thiazoline, 2-isobutyl-4,5-dimethyl-2,5-dihydrothiazole, 2-isobutyl-4,5-dimethyl-3-thiazoline, 2,5-dihydro-4,5-dimethyl-2-2-methylpropyl thiazole, thiazole, 2,5-dihydro-4,5-dimethyl-2-2-methylpropyl, fema no. 3621, 2-isobutyl-4,5-dimethyl thiazoline, 4,5-dimethyl-2-2-methylpropyl-2,5-dihydro-1,3-thiazole, 2,5-dihydro-2-isobutyl-4,5-dimethylthiazole, 3-thiazoline, 4,5-dimethyl-2-isobutyl
PubChem CID 4120025
IUPAC Name 4,5-dimethyl-2-(2-methylpropyl)-2,5-dihydro-1,3-thiazole
SMILES CC(C)CC1SC(C)C(C)=N1

Specifications

Color Yellow
Boiling Point 70°C
Quantity 25 g
Formula Weight 171.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers)
RTECSNumber XJ6642800
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.