Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isobutyl-4-methyl-1,3-dioxolane 98.0+%, TCI America™
SDP

Catalog No. I089625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I089625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I089625ML Supplier TCI America Supplier No. I089625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18433-93-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00238528
InChI Key PDVLTWPJDBXATJ-UHFFFAOYNA-N
Synonym Isovaleraldehyde Propylene Glycol Acetal, 3-Methylbutanal Propylene Glycol Acetal
PubChem CID 86733
IUPAC Name 4-methyl-2-(2-methylpropyl)-1,3-dioxolane
SMILES CC(C)CC1OCC(C)O1

Specifications

Color Colorless
Boiling Point 153°C
Quantity 25 mL
UN Number 1993
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isobutyl-4-methyl-1,3-dioxolane
EINECSNumber (5)-0506
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.