Alfa Aesar™ 2-Isopropylphenol, 98%

Manufacturer: Alfa Aesar™ A1202718

Quantity 50g

 View more versions of this product

Catalog No. AAA1202718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88-69-7
    Molecular Formula C9H12O
    Formula Weight 136.2
    MDL Number MFCD00002224
    UN Number UN2922
    Beilstein 1363322
    Synonym o-Cumenol; o-Hydroxycumene
    Chemical Name or Material 2-Isopropylphenol

    Specifications

    Formula Weight 136.2
    Assay Percent Range 98%
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 88-69-7
    UN Number UN2922
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Melting Point 14° to 16°C
    Quantity 50g
    TSCA TSCA
    Molecular Formula C9H12O
    Beilstein 1363322
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    RTECS Number SL5900000
    Flash Point 88°C (190°F)
    Boiling Point 212° to 214°C
    MDL Number MFCD00002224
    Synonym o-Cumenol; o-Hydroxycumene
    Chemical Name or Material 2-Isopropylphenol
    Assay 98%

    GHS H Statement
    H301-H311-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit