Learn More
2-Isopropylphenyl isocyanate, 97%
Description
2-Isopropylphenyl isocyanate is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Isopropylphenyl isocyanate is used as intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
Specifications
Specifications
| Boiling Point | 66°C to 67°C (5 mmHg) |
| Flash Point | 87°C (188°F) |
| Quantity | 1 g |
| UN Number | UN2206 |
| Beilstein | 3588928 |
| Formula Weight | 161.21 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Isopropylphenyl isocyanate |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H300-H330-H334-H227-H315-H319-H335 Fatal if swallowed. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment is urgent (see label). Rinse mouth. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S. |
| Recommended Storage | 2° to 8°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.