Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-5-methoxybenzimidazole, 99%

Catalog No. AAB2057006
Click to view available options
Quantity:
5 g

CAS: 37052-78-1 | C8H8N2OS | 180.23 g/mol

2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.

Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37052-78-1
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.23
MDL Number MFCD00134581
InChI Key KOFBRZWVWJCLGM-UHFFFAOYSA-N
Synonym 5-methoxy-2-mercaptobenzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-methoxy-1h-benzimidazole, 5-methoxy-1h-benzoimidazole-2-thiol, 1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 5-methoxybenzimidazole-2-thiol
PubChem CID 665603
SMILES COC1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point 255°C to 259°C
Quantity 5 g
Beilstein 3605123
Solubility Information Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water.
IUPAC Name 5-methoxy-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Formula Weight 180.23
Percent Purity 99%
Chemical Name or Material 2-Mercapto-5-methoxybenzimidazole
Hazard Category H302-H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
EINECSNumber 253-326-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.