Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-5-nitrobenzimidazole 98.0+%, TCI America™
SDP

Catalog No. M13255G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. M13255G

Supplier: TCI America M13255G

Chemical Identifiers

CAS 6325-91-3
Molecular Formula C7H5N3O2S
Molecular Weight (g/mol) 195.20
MDL Number MFCD00804727
InChI Key YPXQSGWOGQPLQO-UHFFFAOYSA-N
Synonym 2-mercapto-5-nitrobenzimidazole, 5-nitro-2-benzimidazolethiol, 5-nitro-2-mercaptobenzimidazole, 5-nitro-1h-benzo d imidazole-2-thiol, 2-benzimidazolethiol, 5-nitro, 2-benzimidazolinethione, 5-nitro, 2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro, 6-nitro-1h-benzimidazole-2-thiol, 5-nitro-1h-benzimidazole-2-thiol, 5-nitro-3-hydrobenzimidazole-2-thione
PubChem CID 3717598
IUPAC Name 5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
SMILES [O-][N+](=O)C1=CC=C2NC(=S)NC2=C1

Specifications

Quantity 5 g
Formula Weight 195.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.