Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-6-nitrobenzothiazole 96%, Thermo Scientific™

Catalog No. AC413610050
Click to view available options
:
5g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4845-58-3
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Molecular Formula C7H4N2O2S2
Quantity 5g
MDL Number MFCD00041850
Synonym 2-mercapto-6-nitrobenzothiazole, 6-nitrobenzo d thiazole-2-thiol, 6-nitro-2-mercaptobenzothiazole, 6-nitrobenzo d thiazole-2 3h-thione, 6-nitrobenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-nitro, usaf ek-3991, 6-nitro-1,3-benzothiazole-2-thiol, 2-benzothiazolinethione, 6-nitro, 2-benzothiazolethiol, 6-nitro
InChI Key QPOZGXKWWKLJDK-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC=C2NC(=S)SC2=C1
IUPAC Name 6-nitro-2,3-dihydro-1,3-benzothiazole-2-thione
Molecular Weight (g/mol) 212.24
PubChem CID 947375
Formula Weight 212.25
Percent Purity 96%
Chemical Name or Material 2-Mercapto-6-nitrobenzothiazole, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
RTECSNumber DL6600000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.