Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Mercaptobenzothiazole, 95%
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator

Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 172.0°C to 182.0°C |
Color | Yellow |
Flash Point | 243°C |
Loss on Drying | 0.5% max. (60°C, 3 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Plastic bottle |
Quantity | 50 g |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves. |
EINECSNumber | 205-736-8 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction