Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptobenzothiazole, 95%

Catalog No. p-3758284
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle

CAS: 149-30-4 | C7H5NS2 | 167.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-30-4
Molecular Formula C7H5NS2
Molecular Weight (g/mol) 167.24
MDL Number MFCD00005781
InChI Key YXIWHUQXZSMYRE-UHFFFAOYSA-N
Synonym 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
PubChem CID 697993
ChEBI CHEBI:34292
IUPAC Name 3H-1,3-benzothiazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)S2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 172.0°C to 182.0°C
Color Yellow
Flash Point 243°C
Loss on Drying 0.5% max. (60°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 10 g
Beilstein 27, II, 233
Merck Index 15, 5935
Solubility Information Solubility in water: 0.12g/L (25°C). Other solubilities: soluble in alkalies,soluble in alcohol,acetone,benzene and,chloroform
Formula Weight 167.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Mercaptobenzothiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves.
EINECSNumber 205-736-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.