Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoethanesulfonic acid, sodium salt, MP Biomedicals™

Catalog No. ICN15160210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15160210 10 g
ICN15160201 1 g
ICN15160225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15160210 Supplier MP Biomedicals, Inc Supplier No. 0215160210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19767-45-4 | Formula: C2H5NaO3S2 | Weight: 164.169 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19767-45-4
Molecular Formula C2H5NaO3S2
Molecular Weight (g/mol) 164.169
InChI Key XOGTZOOQQBDUSI-UHFFFAOYSA-M
Synonym mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
PubChem CID 23662354
IUPAC Name sodium;2-sulfanylethanesulfonate
SMILES C(CS(=O)(=O)[O-])S.[Na+]

Specifications

Color White
Assay Percent Range 0.995
Quantity 10 g
Formula Weight 164.2
Physical Form Powder
Chemical Name or Material 2-Mercaptoethanesulfonic acid, sodium salt, For HPLC analysis
Recommended Storage Store at 2° to 8°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.