Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoethyl sulfide, 90%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
3570-55-6
C4H10S3
154.32
KSJBMDCFYZKAFH-UHFFFAOYSA-N
2,2'-thiodiethanethiol,bis 2-mercaptoethyl sulfide,2-mercaptoethyl sulfide,ethanethiol, 2,2'-thiobis,bis mercaptoethyl sulfide,3-thia-1,5-pentanedithiol,2-mercaptoethylsulfide,2,2'-dimercaptodiethyl sulfide,unii-oeu4azc07s,ethanethiol, 2,2'-thiodi
77117
2-(2-sulfanylethylsulfanyl)ethanethiol
C(CSCCS)S
3570-55-6
C4H10S3
154.32
KSJBMDCFYZKAFH-UHFFFAOYSA-N
2,2'-thiodiethanethiol,bis 2-mercaptoethyl sulfide,2-mercaptoethyl sulfide,ethanethiol, 2,2'-thiobis,bis mercaptoethyl sulfide,3-thia-1,5-pentanedithiol,2-mercaptoethylsulfide,2,2'-dimercaptodiethyl sulfide,unii-oeu4azc07s,ethanethiol, 2,2'-thiodi
77117
2-(2-sulfanylethylsulfanyl)ethanethiol
C(CSCCS)S
Refractive Index 1.594
Specific Gravity 1.183
Packaging Glass bottle
Flash Point 90°C
Quantity 50g
Infrared Spectrum Authentic
Boiling Point 135.0°C to 136.0°C (10.0mmHg)
Melting Point -11.0°C
Color Colorless
Density 1.1830g/mL
Assay Percent Range 88 % min. (GC)
Linear Formula (HSCH2CH2)2S
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,toluene,chloroform,,dioxane,carbon tetrachloride
Formula Weight 154.32
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material 2-Mercaptoethyl sulfide
Show More Show Less