Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptonicotinic Acid, TCI America™
SDP

Catalog No. m121325g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Catalog No. M121325G

Supplier: TCI America M121325G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38521-46-9
Molecular Formula C6H5NO2S
Molecular Weight (g/mol) 155.171
MDL Number MFCD00010102
InChI Key WYKHFQKONWMWQM-UHFFFAOYSA-N
Synonym 2-mercaptonicotinic acid, 2-sulfanylpyridine-3-carboxylic acid, 2-mercaptopyridine-3-carboxylic acid, 2-mercapto-nicotinic acid, unii-eu7d859abz, 3-pyridinecarboxylic acid, 1,2-dihydro-2-thioxo, 2-sulfanylnicotinic acid, 1,2-dihydro-2-thioxopyridine-3-carboxylic acid, eu7d859abz, 2-mercapto-3-pyridinecarboxylic acid
PubChem CID 673681
IUPAC Name 2-sulfanylidene-1H-pyridine-3-carboxylic acid
SMILES C1=CNC(=S)C(=C1)C(=O)O

Specifications

Quantity 25 g
Formula Weight 155.17
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptonicotinic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.