Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Tri-n-butyltin chloride, 95%, tech.
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100 g
                            
                        
                            
                                500 g
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| Melting Point | -9.0°C | 
| Color | Colorless to Yellow | 
| Density | 1.2000g/mL | 
| Boiling Point | 171.0°C to 173.0°C (25.0 mmHg) | 
| Flash Point | >112°C | 
| Infrared Spectrum | Authentic | 
| Assay Percent Range | 94% min. (Argentometry) | 
| Packaging | Glass bottle | 
| Refractive Index | 1.4880 to 1.4940 | 
| Linear Formula | [CH3(CH2)3]3SnCl | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Danger | 
| Hazard Statement | GHS H Statement Harmful in contact with skin. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. Causes damage to organs through prolonged or repeated exposure. Causes serious eye irritation. Ca | 
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clo | 
| EINECSNumber | 215-958-7 | 
| RTECSNumber | WH6820000 | 
| TSCA | TSCA | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                