Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
2.5g
25g
100g
Packaging:
Glass bottle
2637-34-5
C5H5NS
111.16
MFCD00006285
WHMDPDGBKYUEMW-UHFFFAOYSA-N
2-mercaptopyridine,2-pyridinethiol,pyridine-2-thiol,2-thiopyridine,pyrid-2-thione,2 1h-pyridinethione,pyridinethione,2-thiopyridone,2-pyridinethione,2-mercapto pyridine
2723698
CHEBI:45223
1H-pyridine-2-thione
C1=CC(=S)NC=C1
2637-34-5
C5H5NS
111.16
MFCD00006285
WHMDPDGBKYUEMW-UHFFFAOYSA-N
2-mercaptopyridine,2-pyridinethiol,pyridine-2-thiol,2-thiopyridine,pyrid-2-thione,2 1h-pyridinethione,pyridinethione,2-thiopyridone,2-pyridinethione,2-mercapto pyridine
2723698
CHEBI:45223
1H-pyridine-2-thione
C1=CC(=S)NC=C1
Melting Point 126.0°C to 130.0°C
Packaging Glass bottle
Flash Point 125°C
Infrared Spectrum Authentic
Quantity 100g
Color Brown to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Iodimetry)
Linear Formula (C5H4N)SH
Beilstein 21,45
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in most common solvents
Formula Weight 111.16
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyridine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-131-9