Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptothiazole, 97%

Catalog No. AAL0705806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0705806 5 g
AAL0705803 1 g
AAL0705814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0705806 Supplier Thermo Scientific Chemicals Supplier No. L0705806
Add to Cart

CAS: 82358-09-6 | C3H3NS2 | 117.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82358-09-6
Molecular Formula C3H3NS2
Molecular Weight (g/mol) 117.18
MDL Number MFCD00022449
InChI Key OCVLSHAVSIYKLI-UHFFFAOYSA-N
Synonym 2-mercaptothiazole, 2 3h-thiazolethione, 2-thiazolethiol, 1,3-thiazole-2-thiol, thiazoline-2-thione, 2-mercapto-1,3-thiazole, 3h-thiazole-2-thione, thiazole-2-thiol, 1,3-thiazole-2 3h-thione, .delta.4-thiazoline-2-thione
PubChem CID 1201387
SMILES S=C1NC=CS1

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Beilstein 635950
IUPAC Name 2,3-dihydro-1,3-thiazole-2-thione
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material 2-Mercaptothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 423-680-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.