Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mesitylenesulfonyl Chloride 99.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
773-64-8
C9H11ClO2S
218.695
MFCD00007434
PVJZBZSCGJAWNG-UHFFFAOYSA-N
2-mesitylenesulfonyl chloride,mesitylenesulfonyl chloride,2,4,6-trimethylbenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 2,4,6-trimethyl,mesitylsulfonyl chloride,2-mesitylenesulfonylchloride,mesitylene-2-sulphonyl chloride,2,4,6-trimethyl-benzenesulfonyl chloride,2,4,6-trimethylphenylsulfonyl chloride,mesitylsulfonylchloride
13046
2,4,6-trimethylbenzenesulfonyl chloride
CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C
773-64-8
C9H11ClO2S
218.695
MFCD00007434
PVJZBZSCGJAWNG-UHFFFAOYSA-N
2-mesitylenesulfonyl chloride,mesitylenesulfonyl chloride,2,4,6-trimethylbenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 2,4,6-trimethyl,mesitylsulfonyl chloride,2-mesitylenesulfonylchloride,mesitylene-2-sulphonyl chloride,2,4,6-trimethyl-benzenesulfonyl chloride,2,4,6-trimethylphenylsulfonyl chloride,mesitylsulfonylchloride
13046
2,4,6-trimethylbenzenesulfonyl chloride
CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C
Color White
Quantity 25g
Melting Point 56°C
Boiling Point 150°C
UN Number 3261
Formula Weight 218.70
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (T)
Chemical Name or Material 2-Mesitylenesulfonyl Chloride

RTECSNumber : DB8930000

TSCA : No

Recommended Storage : Refrigerator