Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mesitylethanol 98.0+%, TCI America™
SDP

Catalog No. M26241G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 6950-92-1
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00020609
InChI Key FQZPTDPHVFTOSY-UHFFFAOYSA-N
Synonym 2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol
PubChem CID 81389
IUPAC Name 2-(2,4,6-trimethylphenyl)ethan-1-ol
SMILES CC1=CC(C)=C(CCO)C(C)=C1

Specifications

Melting Point 80°C
Color White
Boiling Point 152°C
Quantity 1 g
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mesitylethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.