Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mesitylmagnesium bromide, 1M solution in THF, AcroSeal™

Catalog No. ac210731000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC210731000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC210731000 Supplier Thermo Scientific Chemicals Supplier No. 210731000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 2633-66-1 | C9H11BrMg | 223.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2633-66-1 , 109-99-9
Molecular Formula C9H11BrMg
Molecular Weight (g/mol) 223.40
MDL Number MFCD00009663
InChI Key VZMGGVXGYWKQPB-UHFFFAOYSA-M
Synonym 2-mesitylmagnesium bromide, mesitylmagnesium bromide, 2,4,6-trimethylphenylmagnesium bromide, 2-mesitylmagnesium bromide solution, mesmgbr, yxvsitsudrgill-uhfffaoysa-m, magnesium,bromo 2,4,6-trimethylphenyl, 2-mesitylmagnesium bromide solution, 1.0 m in thf, 2,4,6-trimethylphenylmagnesium bromide, 1m in 2-methf, 2-mesitylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
PubChem CID 5098599
IUPAC Name bromo(3,4,5-trimethylphenyl)magnesium
SMILES CC1=CC([Mg]Br)=CC(C)=C1C

Specifications

Color Yellow to Brown
CAS Min % 21.12, 75.55
CAS Max % 24.45, 78.88
Physical Form Solution
Quantity 100 mL
Linear Formula (CH3)3C6H2MgBr
Flash Point −17°C
Formula Weight 223.39
Concentration or Composition (by Analyte or Components) 0.95 to 1.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.005
Density 1.0050g/mL
Chemical Name or Material 2-Mesitylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.
May cause drowsiness or dizziness.<b
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.