Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-4-methylphenol, 99%

Catalog No. p-157447
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 93-51-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-51-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002378
InChI Key PETRWTHZSKVLRE-UHFFFAOYSA-N
Synonym creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene
PubChem CID 7144
IUPAC Name 2-methoxy-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 2.0°C
Color Yellow
Density 1.0900g/mL
Boiling Point 221.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5360 to 1.5380
Quantity 25 g
Specific Gravity 1.09
Merck Index 15, 2559
Solubility Information Solubility in water: insoluble
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methoxy-4-methylphenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 202-252-9
RTECSNumber GP1755000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.