Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxy-4-nitroaniline, 99%

Click to view available options
Quantity:
25g
100g
500g
97-52-9
C7H8N2O3
168.152
MFCD00007363
GVBHRNIWBGTNQA-UHFFFAOYSA-N
fast red b base,benzenamine, 2-methoxy-4-nitro,4-nitro-o-anisidine,fast red base b,fast red b,red b base,diazo fast red b,2-amino-5-nitroanisole,azoamine pink o,fast red 5na base
7337
2-methoxy-4-nitroaniline
COC1=C(C=CC(=C1)[N+](=O)[O-])N
97-52-9
C7H8N2O3
168.152
MFCD00007363
GVBHRNIWBGTNQA-UHFFFAOYSA-N
fast red b base,benzenamine, 2-methoxy-4-nitro,4-nitro-o-anisidine,fast red base b,fast red b,red b base,diazo fast red b,2-amino-5-nitroanisole,azoamine pink o,fast red 5na base
7337
2-methoxy-4-nitroaniline
COC1=C(C=CC(=C1)[N+](=O)[O-])N
Melting Point 138°C to 142°C
Quantity 25g
Beilstein 879619
Formula Weight 168.15
Percent Purity 99%
Chemical Name or Material 2-Methoxy-4-nitroaniline

GHS H Statement
H301-H311-H341-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BZ7170000

TSCA : TSCA