Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-methylphenyl isocyanate, 98%, Thermo Scientific™

Catalog No. AC441770050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441770050 5 g
AC441770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441770050 Supplier Thermo Scientific Chemicals Supplier No. 441770050

Chemical Identifiers

CAS 59741-04-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
InChI Key IDOHLSFNBKNRRJ-UHFFFAOYSA-N
Synonym 2-methoxy-5-methylphenyl isocyanate, 2-methoxy-5-methylphenylisocyanate, 2-isocyanato-1-methoxy-4-methyl-benzene, benzene, 2-isocyanato-1-methoxy-4-methyl, acmc-20anrw, 2-methoxy-5-methylbenzenisocyanate, 2-methoxy-5-methylpheny lisocyanate, 2-isocyanato-1-methyloxy-4-methylbenzene, benzene, 2-isocyanato-1-methoxy-4-methyl-9ci
PubChem CID 4141520
IUPAC Name 2-isocyanato-1-methoxy-4-methylbenzene
SMILES CC1=CC(=C(C=C1)OC)N=C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 25.0°C to 26.0°C
Color Colorless
Density 1.1120g/mL
Boiling Point 240.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.112
Formula Weight 163.18
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Methoxy-5-methylphenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reactio
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.