Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-sulfamoylbenzoic acid, 97%

Catalog No. AAB2369809
Click to view available options
Quantity:
10 g
50 g

CAS: 22117-85-7 | C8H9NO5S | 231.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22117-85-7
Molecular Formula C8H9NO5S
Molecular Weight (g/mol) 231.222
MDL Number MFCD00129997
InChI Key SQAILWDRVDGLGY-UHFFFAOYSA-N
Synonym 2-methoxy-5-aminosulfonylbenzoic acid, benzoic acid, 5-aminosulfonyl-2-methoxy, 5-sulphamoyl-o-anisic acid, 2-methoxy-5-sulfonylbenzoic acid, sulpiride impurity d, acmc-209fsc, 2-methoxy-5-sulfamoylbenzoicacid, 2-methoxy-5-sulphamoylbenzoic acid, 2-methoxy-5-sulfamoyl benzoic acid, 2-methoxy-5-sulfamoyl-benzoic acid
PubChem CID 89601
IUPAC Name 2-methoxy-5-sulfamoylbenzoic acid
SMILES COC1=C(C=C(C=C1)S(=O)(=O)N)C(=O)O

Specifications

Quantity 10 g
Formula Weight 232.23
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-sulfamoylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-789-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.