Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzhydrazide, 98+%

Catalog No. AAA1361918
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1361918 50 g
AAA1361909 10 g
AAA1361930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1361918 Supplier Thermo Scientific Chemicals Supplier No. A1361918
Add to Cart

CAS: 7466-54-8 | C8H10N2O2 | 166.18 g/mol

2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7466-54-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014755
InChI Key QMGXWNSSMGAHCA-UHFFFAOYSA-N
Synonym 2-methoxybenzhydrazide, 2-methoxybenzoic acid hydrazide, o-methoxybenzohydrazide, o-methoxybenzoylhydrazine, 2-methoxybenzoylhydrazide, 2-methoxybenzoylhydrazine, o-anisohydrazide, hydrazine, 2-methoxybenzoyl, o-methoxybenzoylhydrazide, o-methoxybenzoic acid hydrazide
PubChem CID 24051
IUPAC Name 2-methoxybenzohydrazide
SMILES COC1=CC=CC=C1C(=O)NN

Specifications

Melting Point 78°C to 82°C
Quantity 50 g
Beilstein 2208367
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybenzhydrazide
EINECSNumber 231-260-5
RTECSNumber BZ4700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.