Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6609-56-9
C8H7NO
133.15
MFCD00001783
FSTPMFASNVISBU-UHFFFAOYSA-N
2-cyanoanisole,o-methoxybenzonitrile,2-methoxy-benzonitrile,benzonitrile, 2-methoxy,2-methoxybenzenecarbonitrile,o-anisolnitrile,o-anisonitrile,methoxybenzonitrile,2-methxybenzonitrile,o-methyoxybenzonitrile
81086
2-methoxybenzonitrile
COC1=CC=CC=C1C#N
6609-56-9
C8H7NO
133.15
MFCD00001783
FSTPMFASNVISBU-UHFFFAOYSA-N
2-cyanoanisole,o-methoxybenzonitrile,2-methoxy-benzonitrile,benzonitrile, 2-methoxy,2-methoxybenzenecarbonitrile,o-anisolnitrile,o-anisonitrile,methoxybenzonitrile,2-methxybenzonitrile,o-methyoxybenzonitrile
81086
2-methoxybenzonitrile
COC1=CC=CC=C1C#N
Boiling Point 256°C
Flash Point 113°C
Melting Point 21°C to 22°C
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 25g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 133.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Methoxybenzonitrile
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

EINECSNumber : 229-559-