Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzylamine, 98+%

Catalog No. AAL0227314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0227314 25 g
AAL0227322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0227314 Supplier Thermo Scientific Chemicals Supplier No. L0227314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 6850-57-3 | C8H11NO | 137.182 g/mol

2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6850-57-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00008110
InChI Key PXJACNDVRNAFHD-UHFFFAOYSA-N
Synonym 2-methoxybenzylamine, 2-methoxyphenyl methanamine, o-methoxybenzylamine, benzenemethanamine, 2-methoxy, 2-methoxy-benzylamine, 2-methoxyphenyl methylamine, 1-2-methoxyphenyl methanamine, 2-methyoxybenzylamine, 2-methoxy benzylamine, 2-methoxybenzyl amine
PubChem CID 81292
IUPAC Name (2-methoxyphenyl)methanamine
SMILES COC1=CC=CC=C1CN

Specifications

Density 1.057
Boiling Point 228°C to 229°C
Flash Point 100°C (212°F)
Refractive Index 1.548
Quantity 25 g
UN Number UN2735
Beilstein 1100448
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybenzylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 229-941-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.