Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxycyclohexanone 95.0+%, TCI America™
SDP

Catalog No. M117125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M117125G 25 g
M11715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M117125G Supplier TCI America Supplier No. M117125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7429-44-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009610
InChI Key JYJURPHZXCLFDX-UHFFFAOYSA-N
PubChem CID 110998
IUPAC Name 2-methoxycyclohexan-1-one
SMILES COC1CCCCC1=O

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 g
Formula Weight 128.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxycyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.