Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper(II) acetate, 99%, pure, anhydrous
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Oxidation reagent
Specifications
Specifications
| Melting Point | 115.0°C |
| Color | Blue to Green |
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| Assay Percent Range | 99% |
| Linear Formula | (CH3CO2)2Cu |
| Fieser | 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 15,99 |
| Merck Index | 15,2614 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 181.64 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if pre |
| EINECSNumber | 205-553-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction