Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, ACS, 99%3, Spectrum™ Chemical
SDP

Catalog No. 18608225 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-225 20 L Amber Glass Bottle
18-608-227 500 mL Amber Glass Bottle
18-608-224 4 L Amber Glass Bottle
18-608-226 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-608-225 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M122120LTMT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
IUPAC Name 2-methoxyethan-1-ol
SMILES COCCO

Specifications

CAS Max % 1
Boiling Point 125°C (257°F)
Packaging Amber Glass Bottle
Quantity 20 L
Percent Purity 0.993
Grade ACS
Titratable Acid 0.002 meq/g
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.