Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, puriss. p.a., ACS Reagent, ≥99.5% (GC), Solstice

Catalog No. 60046257 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-257 1 L Poly Bottle
60-046-258 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-046-257 Supplier Solstice Supplier No. 334571L
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point -85°C
Density 0.966-0.968g/mL at 20°C, 0.965g/mL at 25°C
Boiling Point 124°C
Explosion Limit 0.245
Flash Point 40°C (104°F)
Ignition Point 548°F
Packaging Poly Bottle
Refractive Index n20/D 1.402
Linear Formula CH3OCH2CH2OH
Quantity 1 L
UN Number UN1188
Residue after Evaporation 0 to 0.002%
Beilstein 1731074
Vapor Pressure 6.17mmHg (20°C)
Formula Weight 76.09g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
Hazard Statement P201-P210-P260-P280-P308 + P311-P370 + P378
DOTInformation class 3 - PG 3 - Ethylene glycol monomethyl ether
RTECSNumber 203-713-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.