Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethyl Methanesulfonate 97.0+%, TCI America™
SDP

Catalog No. M144625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M144625G 25 g
M14465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M144625G Supplier TCI America Supplier No. M144625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16427-44-4
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD02656401
InChI Key BCKAHDGFNHDQST-UHFFFAOYSA-N
Synonym Methanesulfonic Acid 2-Methoxyethyl Ester
PubChem CID 146293
IUPAC Name 2-methoxyethyl methanesulfonate
SMILES COCCOS(=O)(=O)C

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 g
Formula Weight 154.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.