Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyhydroquinone, 97%

Catalog No. AAA1284718
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1284718 50 g
AAA1284709 10 g
AAA1284730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1284718 Supplier Thermo Scientific Chemicals Supplier No. A1284718
Add to Cart

CAS: 824-46-4 | C7H8O3 | 140.138 g/mol

It finds its application in the formation of coniferyl alcohol from ferulic acid by the white rot fungus Trametes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the formation of coniferyl alcohol from ferulic acid by the white rot fungus Trametes.

Solubility
Slightly soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-46-4
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00013971
InChI Key LAQYHRQFABOIFD-UHFFFAOYSA-N
Synonym 2-methoxyhydroquinone, methoxyhydroquinone, 1,4-benzenediol, 2-methoxy, 1,4-dihydroxy-2-methoxybenzene, unii-2hi6hnr5u1, 2hi6hnr5u1, o-methoxyhydroquinone, 2,5-dihydroxyanisole, 2-mohydrop, acmc-209pp0
PubChem CID 69988
IUPAC Name 2-methoxybenzene-1,4-diol
SMILES COC1=C(C=CC(=C1)O)O

Specifications

Melting Point 87°C to 91°C
Quantity 50 g
Beilstein 2045285
Solubility Information Slightly soluble in water. Solubility in methanol is almost transparent.
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 2-Methoxyhydroquinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-530-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.